4-Chloro-2-methylquinazoline 95%

Supplier: Ambeed
Warning

A115508-10G A115508-25G A115508-1G A115508-5G A115508-250MG
103798-578EA 22 USD
103798-578 103824-436 76781-372 76781-374 77311-252 77311-254 77592-262
4-Chloro-2-methylquinazoline 95%
4-Chloro-2-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.6183 g/mol
Boiling Pt: 211.2 °C (760 mm Hg)
Melting Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05666292
CAS Number: 6484-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR