4-Chloro-o-toluic acid 98%

Supplier: Ambeed
Warning

A156464-100G A156464-10G A156464-500G A156464-1G A156464-25G A156464-5G
103804-886EA 6 USD
103804-886 103843-088 77286-280 77286-282 77286-284 77300-186 77300-188 77300-190
4-Chloro-o-toluic acid 98%
4-Chloro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 300.3 °C (760 mm Hg)
Melting Pt: 167…171 °C
Flash Pt: 135.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00045852
CAS Number: 7499-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR