4-Chloro-2-methyl-7-azaindole 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine
A307921-100MG
A307921-10G
A307921-250MG
A307921-1G
103814-536EA
64
USD
103814-536
103835-712
76875-736
76875-738
76875-740
77305-974
4-Chloro-2-methyl-7-azaindole 95%
4-Chloro-2-methyl-7-azaindole
Precaución: for Research Use Only
Formula:
C₈H₇ClN₂ MW: 166.608 g/mol Melting Pt: 173…178 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12924600 CAS Number: 307951-53-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...