4-Chloro-o-anisidine 97%

Supplier: Ambeed
Warning

A142573-10G A142573-250MG A142573-1G A142573-5G
103803-188EA 8 USD
103803-188 77150-132 77150-134 77316-414 77316-416
4-Chloro-o-anisidine 97%
4-Chloro-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
Boiling Pt: 260 °C (760 mm Hg)
Flash Pt: 108.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06801386
CAS Number: 93-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR