4-Chloro-2-iodopyridine 95%

Supplier: Ambeed
Warning

A223805-100MG A223805-250MG A223805-5G
103819-066EA 111 USD
103819-066 103849-948 76814-960 76814-962 77088-574
4-Chloro-2-iodopyridine 95%
4-Chloro-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIN
MW: 239.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913885
CAS Number: 22918-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR