4-Chloro-2-iodophenol 98%

Supplier: Ambeed
Warning

A175204-1G A175204-25G A175204-5G A175204-10G A175204-250MG
103807-008EA 20 USD
103807-008 103845-314 77082-638 77082-640 77082-642 77286-992 77286-994
4-Chloro-2-iodophenol 98%
4-Chloro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
Boiling Pt: 225.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08235248
CAS Number: 71643-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR