4-Chloro-2-iodobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A287198-1G
A287198-5G
A287198-100G
A287198-10G
A287198-25G
103813-738EA
6
USD
103813-738
103834-878
76837-416
76837-418
77289-748
77289-750
77305-510
4-Chloro-2-iodobenzoic acid 98%
4-Chloro-2-iodobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄ClIO₂ MW: 282.46 g/mol Boiling Pt: 343.8 °C (760 mm Hg) Melting Pt: 163…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060676 CAS Number: 13421-13-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...