4-Chlorosalicylaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-2-hydroxybenzaldehyde
A100481-100G
A100481-10G
A100481-250MG
A100481-25G
A100481-1G
A100481-5G
103795-466EA
13
USD
103795-466
103821-160
77135-438
77135-440
77135-442
77135-444
77296-710
77296-712
4-Chlorosalicylaldehyde 98%
4-Chlorosalicylaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅ClO₂ MW: 156.5664 g/mol Boiling Pt: 229.1 °C (760 mm Hg) Melting Pt: 45…50 °C Flash Pt: 92.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06252499 CAS Number: 2420-26-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...