4-Chloro-2-formylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-formylbenzeneboronic acid

A403216-1G A403216-5G A403216-10G A403216-250MG A403216-25G
103827-088EA 23 USD
103827-088 103838-128 76841-598 76841-600 77291-182 77291-184 77748-810
4-Chloro-2-formylphenylboronic acid 98%
4-Chloro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClO₃
MW: 184.39 g/mol
Boiling Pt: 371.8 °C (760 mm Hg)
Melting Pt: 132…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689492
CAS Number: 913835-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR