4-Chloro-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A122928-100G A122928-10G A122928-25G A122928-500G A122928-1G A122928-5G
103800-310EA 7 USD
103800-310 76971-646 76971-648 76971-650 76971-652 77312-426 77312-428
4-Chloro-2-fluoropyridine 98%
4-Chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃ClFN
MW: 131.53 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04112503
CAS Number: 34941-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR