(4-Chloro-2-fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-fluorophenylacetic acid

A145383-100G A145383-10G A145383-25G A145383-1G A145383-5G
103803-528EA 5 USD
103803-528 103841-616 77285-908 77285-910 77285-912 77316-902 77316-904
(4-Chloro-2-fluorophenyl)acetic acid 98%
(4-Chloro-2-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
Boiling Pt: 287.3 °C (760 mm Hg)
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD03095402
CAS Number: 194240-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR