4-Chloro-2,6-diphenylpyrimidine 95%

Supplier: Ambeed
Warning

A232700-25G A232700-5G A232700-10G A232700-1G
103810-844EA 7 USD
103810-844 103850-598 77089-528 77089-530 77288-450 77303-942
4-Chloro-2,6-diphenylpyrimidine 95%
4-Chloro-2,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₁₆H₁₁ClN₂
MW: 266.73 g/mol
Boiling Pt: 339.2 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 189.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00234890
CAS Number: 29509-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR