4-Chloro-2,6-dimethylquinazoline 95%

Supplier: Ambeed
Warning

A101655-100MG A101655-250MG
103796-068EA 60 USD
103796-068 103821-764 77008-340 77008-342
4-Chloro-2,6-dimethylquinazoline 95%
4-Chloro-2,6-dimethylquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂
MW: 192.65 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22397500
CAS Number: 1429782-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR