4-Chloro-2,6-dimethylpyrimidine 98%

Supplier: Ambeed
Warning

A156749-100G A156749-10G A156749-250MG A156749-1G A156749-25G A156749-5G
103804-920EA 6 USD
103804-920 103843-122 77110-514 77110-516 77110-518 77300-212 77318-920 77318-922
4-Chloro-2,6-dimethylpyrimidine 98%
4-Chloro-2,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.03 g/mol
Boiling Pt: 197.5 °C (760 mm Hg)
Flash Pt: 91.285 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234197
CAS Number: 4472-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR