4-Chloro-2,6-dimethylpyridine 98%

Supplier: Ambeed
Warning

A183886-10G A183886-1G A183886-25G
103815-680EA 20 USD
103815-680 103846-242 76985-818 77083-850 77083-852
4-Chloro-2,6-dimethylpyridine 98%
4-Chloro-2,6-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
Boiling Pt: 179.8 °C (760 mm Hg)
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646174
CAS Number: 3512-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR