4-Chloro-2,6-dimethyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

A232863-1G A232863-25G A232863-5G A232863-10G
103810-860EA 11 USD
103810-860 103850-616 77089-554 77089-556 77089-558 77288-452
4-Chloro-2,6-dimethyl-3-nitropyridine 98%
4-Chloro-2,6-dimethyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.595 g/mol
Boiling Pt: 270.3 °C (760 mm Hg)
Melting Pt: 70…73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08543468
CAS Number: 15513-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR