4-Chloro-2,6-difluoroaniline 97%
Supplier: Ambeed
|
Warning
|
A168866-1G
A168866-25G
A168866-5G
A168866-100G
A168866-10G
103806-326EA
15
USD
103806-326
103844-582
77081-648
77081-650
77081-652
77286-726
77286-728
4-Chloro-2,6-difluoroaniline 97%
4-Chloro-2,6-difluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₄ClF₂N MW: 163.55 g/mol Boiling Pt: 180.9 °C (760 mm Hg) Melting Pt: 47…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426173 CAS Number: 69411-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...