4-Chloro-2,5-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A475204-250MG A475204-25G A475204-1G A475204-100G
103828-070EA 7 USD
103828-070 103839-142 76774-324 76774-326 76843-070 77878-242
4-Chloro-2,5-difluorobenzonitrile 97%
4-Chloro-2,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₂N
Boiling Pt: 206.8 °C (760 mm Hg)
Flash Pt: 78.9 °C
Storage Temperature: Ambient
MDL Number: MFCD06659464
CAS Number: 135748-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR