4-Chloro-2,5-difluorobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A167785-10G
A167785-250MG
A167785-1G
A167785-25G
103806-222EA
81
USD
103806-222
103844-482
76769-006
76769-008
77081-514
77081-516
4-Chloro-2,5-difluorobenzaldehyde 98%
4-Chloro-2,5-difluorobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O MW: 176.55 g/mol Melting Pt: 42…44 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07782051 CAS Number: 879093-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...