4-Chloro-2,2'-bipyridine 95%

Supplier: Ambeed
Warning

A270810-1G A270810-5G A270810-250MG
103813-016EA 13 USD
103813-016 103852-922 77092-692 77092-694 77289-444
4-Chloro-2,2'-bipyridine 95%
4-Chloro-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
MW: 190.63 g/mol
Boiling Pt: 309.768 °C (760 mm Hg)
Melting Pt: 84…85 °C
Flash Pt: 170.077 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11112152
CAS Number: 14162-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR