4'-Chloro-2,2':6',2''-terpyridine 97%

Supplier: Ambeed
Warning

A212007-10G A212007-250MG A212007-1G A212007-5G
103818-156EA 32 USD
103818-156 103848-852 76785-710 76785-712 77087-292 77087-294
4'-Chloro-2,2':6',2''-terpyridine 97%
4'-Chloro-2,2':6',2''-terpyridine

Precaución: for Research Use Only

Formula: C₁₅H₁₀ClN₃
MW: 267.716 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00191930
CAS Number: 128143-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR