4-Chloro-2-(trifluoromethyl)phenol 95%

Supplier: Ambeed
Warning

A115357-10G A115357-250MG A115357-25G A115357-1G A115357-100MG
103798-556EA 33 USD
103798-556 103824-410 76869-976 76869-978 76869-980 77311-226 77882-948
4-Chloro-2-(trifluoromethyl)phenol 95%
4-Chloro-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00060673
CAS Number: 53903-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR