4-Chloro-2-(trifluoromethoxy)aniline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-2-(trifluoromethoxy)phenylamine
A152004-100MG
A152004-10G
A152004-250MG
A152004-5G
103804-338EA
26
USD
103804-338
103842-488
77286-142
77286-144
77286-146
77318-088
4-Chloro-2-(trifluoromethoxy)aniline 97%
4-Chloro-2-(trifluoromethoxy)aniline
Precaución: for Research Use Only
Formula:
C₇H₅ClF₃NO Boiling Pt: 212 °C (760 mm Hg) Flash Pt: 72.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153102 CAS Number: 175205-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...