4-Chloro-2-(methylsulfanyl)pyrimidine-5-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-(methylthio)pyrimidine-5-carbonitrile

A602225-10G A602225-250MG A602225-1G A602225-25G
103828-984EA 16 USD
103828-984 103840-124 77005-398 77005-400 77307-920 77673-622
4-Chloro-2-(methylsulfanyl)pyrimidine-5-carbonitrile 98%
4-Chloro-2-(methylsulfanyl)pyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃S
Boiling Pt: 340.7 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD00507261
CAS Number: 33089-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR