4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Danger

A509409-100MG A509409-250MG
103828-370EA 152 USD
103828-370 103839-470 76790-006 76790-008
4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Chloro-2-(methylthio)-7H-pyrrolo[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃S
MW: 199.6606 g/mol
Boiling Pt: 433.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09025769
CAS Number: 57564-94-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR