4-Chloro-2-(chloromethyl)pyridine hydrochloride 97%

Supplier: Ambeed
Danger

A277208-1G A277208-250MG A277208-5G
103813-340EA 105 USD
103813-340 77093-162 77289-568 77289-570
4-Chloro-2-(chloromethyl)pyridine hydrochloride 97%
4-Chloro-2-(chloromethyl)pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N·ClH
MW: 198.478 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08234868
CAS Number: 119396-04-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR