4-Chloro-1-naphthoic acid 98%

Supplier: Ambeed
Warning

A453840-100MG A453840-5G A453840-250MG
103827-816EA 48 USD
103827-816 76829-748 77684-244 77684-243
4-Chloro-1-naphthoic acid 98%
4-Chloro-1-naphthoic acid

Precaución: For research use only.

Formula: C₁₁H₇ClO₂
MW: 206.63 g/mol
Melting Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD18252916
CAS Number: 1013-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR