4-Chloro-1-methylpiperidine 97%

Supplier: Ambeed
Warning

A101901-100G A101901-10G A101901-25G A101901-500G
103821-898EA 5 USD
103821-898 77160-226 77160-228 77160-230 77160-232
4-Chloro-1-methylpiperidine 97%
4-Chloro-1-methylpiperidine

Precaución: for Research Use Only

Formula: C₆H₁₂ClN
MW: 133.6192 g/mol
Boiling Pt: 202.4 °C (760 mm Hg)
Flash Pt: 76.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00044489
CAS Number: 5570-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR