4-Chloro-1-methyl-1H-pyrazol-5-amine 98%

Supplier: Ambeed
Warning

A205162-100MG A205162-250MG
103817-606EA 241 USD
103817-606 103848-272 77890-236 77890-237
4-Chloro-1-methyl-1H-pyrazol-5-amine 98%
4-Chloro-1-methyl-1H-pyrazol-5-amine

Precaución: Keep in a dark place, inert atmosphere, room temperature

Formula: C₄H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD19214209
CAS Number: 105675-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR