4-Chloro-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-5-azaindole

A278896-1G A278896-25G A278896-250MG
103813-438EA 28 USD
103813-438 103853-382 77093-310 77093-312 77289-596
4-Chloro-1H-pyrrolo[3,2-c]pyridine 95%
4-Chloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781160
CAS Number: 60290-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR