4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Chloro-7-azaindole-5-carboxylic acid
A140337-100MG
A140337-10G
A140337-250MG
A140337-1G
103802-856EA
111
USD
103802-856
77027-492
77027-494
77027-496
77315-974
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid 97%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅ClN₂O₂ MW: 196.5900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10574983 CAS Number: 920966-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...