4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%

Supplier: Ambeed
Warning

A128484-250MG A128484-25G A128484-5G
103801-140EA 27 USD
103801-140 77285-198 77313-458 77313-460
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MW: 154.5571 g/mol
Boiling Pt: 348.3 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD03030404
CAS Number: 5399-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR