4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Amino-4-chloro-1H-pyrazolo[3,4-d]pyrimidine
A102544-100MG
A102544-10G
A102544-250MG
A102544-1G
103796-386EA
19
USD
103796-386
103822-082
77282-942
77282-944
77282-946
77308-962
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine 97%
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
Precaución: for Research Use Only
Formula:
C₅H₄ClN₅ MW: 169.5700 g/mol Melting Pt: 210 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08059805 CAS Number: 100644-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...