4-Chloro-1H-pyrazolo[3,4-b]pyridine 98%

Supplier: Ambeed
Warning

A121702-100MG A121702-10G A121702-250MG A121702-25G A121702-1G A121702-5G
103800-114EA 17 USD
103800-114 76781-798 76781-800 76781-802 76781-804 77312-242 77312-244
4-Chloro-1H-pyrazolo[3,4-b]pyridine 98%
4-Chloro-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
Boiling Pt: 308.2 °C (760 mm Hg)
Flash Pt: 169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11846313
CAS Number: 29274-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR