4-Chloro-1H-indole-2-carbonitrile 95%

Supplier: Ambeed
Warning

A105135-100MG A105135-250MG
103796-844EA 158 USD
103796-844 76821-516 76821-518
4-Chloro-1H-indole-2-carbonitrile 95%
4-Chloro-1H-indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅ClN₂
MW: 176.602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135759
CAS Number: 4404-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR