4-Chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A316901-1G A316901-5G A316901-10G A316901-250MG
103825-120EA 23 USD
103825-120 103836-052 76838-894 76838-896 77290-244 77290-246
4-Chloro-1H-indazole 98%
4-Chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00211065
CAS Number: 13096-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR