4-Chloro-1H-indazol-5-ol 98%

Supplier: Ambeed
Warning

A127215-100MG A127215-250MG A127215-1G A127215-5G
103800-934EA 188 USD
103800-934 77009-728 77009-730 77298-192 77298-194
4-Chloro-1H-indazol-5-ol 98%
4-Chloro-1H-indazol-5-ol

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659073
CAS Number: 478834-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR