4-Chloro-1,1-dimethoxybutane 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Chlorobutyraldehyde dimethyl Acetal, 4-Chloro-1,1-dimethoxybutane
A715878-1G
A715878-5G
A715878-10G
103840-708EA
20
USD
103840-708
76844-922
76844-924
77006-068
4-Chloro-1,1-dimethoxybutane 95%
4-Chloro-1,1-dimethoxybutane
Precaución: for Research Use Only
Formula:
C₆H₁₃ClO₂ Boiling Pt: 168.3 °C (760 mm Hg) Flash Pt: 43 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09032953 CAS Number: 29882-07-3 UN: 1993 ADR: 3,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...