4'-Chlorobiphenyl-4-carboxaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
[1,1'-Biphenyl]-4-carboxaldehyde, 4'-chloro-, 4'-Chlorobiphenyl-4-carbaldehyde, 4'-Chloro-4-biphenylcarbaldehyde, 4'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde
A182934-10G
A182934-1G
A182934-25G
A182934-5G
103815-588EA
15
USD
103815-588
103846-150
76720-966
77083-728
77083-730
77083-732
4'-Chlorobiphenyl-4-carboxaldehyde 97%
4'-Chlorobiphenyl-4-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₁₃H₉ClO MW: 216.67 g/mol Boiling Pt: 354.3±25.0 °C (760 mm Hg) Melting Pt: 114…115 °C Flash Pt: 183.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01631911 CAS Number: 80565-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...