2-Amino-4'-chlorobiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4'-Chloro-[1,1'-biphenyl]-2-amine

A128577-100G A128577-10G A128577-250MG A128577-1G A128577-25G A128577-5G
103801-158EA 7 USD
103801-158 76767-038 76767-040 76767-042 77313-488 77313-490 77313-492
2-Amino-4'-chlorobiphenyl 97%
2-Amino-4'-chlorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClN
MW: 203.67 g/mol
Boiling Pt: 334.603 °C (760 mm Hg)
Melting Pt: 53…57 °C
Flash Pt: 156.163 °C
Storage Temperature: Ambient
MDL Number: MFCD03424659
CAS Number: 1204-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR