4-Chloro-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-(2-trifluoromethyl)pyridine

A766351-1G A766351-25G A766351-5G A766351-10G A766351-250MG
103840-914EA 16 USD
103840-914 76845-146 76845-148 76845-150 77281-242 77281-244
4-Chloro-2-(trifluoromethyl)pyridine 98%
4-Chloro-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃N
MW: 181.542 g/mol
Boiling Pt: 137.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07368047
CAS Number: 131748-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR