4-Cbz-aminopiperidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Benzyloxycarbonylaminopiperidine, 4-Cbz-Aminopiperidine

A331228-1G A331228-25G A331228-5G A331228-250MG A331228-10G
103825-576EA 10 USD
103825-576 103836-540 76839-528 76839-530 76839-532 77290-450 77748-650
4-Cbz-aminopiperidine 97%
4-Cbz-aminopiperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.2942 g/mol
Boiling Pt: 396.3 °C (760 mm Hg)
Melting Pt: 80…83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038571
CAS Number: 182223-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR