(+/-)-Terpinen-4-ol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Carvomenthenol, (+/-)-4-Hydroxy-4-isopropyl-1-methyl-1-cyclohexene, (+/-)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (+/-)-p-Menth-1-en-4-ol, Terpinen-4-ol, 1-Isopropyl-4-methylcyclohex-3-en-1-ol
A637955-10G
A637955-500G
A637955-25G
A637955-5G
A637955-100G
A637955-1000G
103840-324EA
5
USD
103840-324
77242-950
77242-952
77308-032
77308-034
77594-956
77617-692
(+/-)-Terpinen-4-ol 95%
(+/-)-Terpinen-4-ol
Precaución: for Research Use Only
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 88…90 °C (760 mm Hg) Flash Pt: 82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001562 CAS Number: 562-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...