4-Carboxyphenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Carboxyphenyl)acetic acid, 4-(Carboxymethyl)benzoic acid

A110635-100G A110635-25G A110635-1G A110635-5G A110635-10G
103797-698EA 6 USD
103797-698 103823-506 77093-992 77093-994 77310-308 77310-310 77592-182
4-Carboxyphenylacetic acid 97%
4-Carboxyphenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
Boiling Pt: 402.5 °C (760 mm Hg)
Flash Pt: 211.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01074834
CAS Number: 501-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR