1-Butyl-4-ethynylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 4-Butylphenylacetylene

A192919-100G A192919-10G A192919-25G A192919-5G
103847-102EA 11 USD
103847-102 76721-170 76721-172 77084-994 77084-996
1-Butyl-4-ethynylbenzene 97%
1-Butyl-4-ethynylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄
Boiling Pt: 70…71 °C (3 mm Hg)
Flash Pt: 101.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00173885
CAS Number: 79887-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR