4'-Butyl-[1,1'-biphenyl]-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A341431-250MG A341431-25G A341431-1G A341431-5G
103825-832EA 25 USD
103825-832 103836-808 77290-528 77290-530 77306-534 77306-536
4'-Butyl-[1,1'-biphenyl]-4-carboxylic acid 98%
4'-Butyl-[1,1'-biphenyl]-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₂
MW: 254.32 g/mol
Boiling Pt: 413.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00222812
CAS Number: 59662-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR