4-Butyl-4'-cyanobiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Butyl-[1,1'-biphenyl]-4-carbonitrile

A679777-5G A679777-10G A679777-250MG A679777-25G
103840-536EA 29 USD
103840-536 76844-734 77035-660 77035-662 77308-116
4-Butyl-4'-cyanobiphenyl 98%
4-Butyl-4'-cyanobiphenyl

Precaución: for Research Use Only

Formula: C₁₇H₁₇N
MW: 235.33 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00506009
CAS Number: 52709-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR