4-Butoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A151391-100G A151391-25G A151391-5G
103842-404EA 5 USD
103842-404 76798-666 77317-964 77317-966
4-Butoxybenzaldehyde 98%
4-Butoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 285 °C at N/A mmHg
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003389
CAS Number: 5736-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR