4-Bromothiophene-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-carboxythiophene, 4-Bromo-3-thenoic acid

A207615-1G A207615-25G A207615-5G A207615-100G A207615-10G A207615-250MG
103817-810EA 16 USD
103817-810 103848-492 77086-776 77086-778 77086-780 77287-836 77287-838 77287-840
4-Bromothiophene-3-carboxylic acid 98%
4-Bromothiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₂S
MW: 207.045 g/mol
Boiling Pt: 318 °C (760 mm Hg)
Melting Pt: 157…159 °C
Flash Pt: 146.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01233020
CAS Number: 16694-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR