4-Bromothiophene-3-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A250352-10G
A250352-250MG
A250352-1G
A250352-25G
A250352-5G
103811-920EA
47
USD
103811-920
103851-760
77276-326
77276-328
77304-520
77304-522
77304-524
4-Bromothiophene-3-carbaldehyde 97%
4-Bromothiophene-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₅H₃BrOS MW: 191.05 g/mol Boiling Pt: 237.9 °C (760 mm Hg) Flash Pt: 97.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318960 CAS Number: 18791-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...